Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab19
Entrez Gene ID
550000
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
rab19b
Chromosome Location
Chr.3: 208919 - 213070 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017246.2 NP_001017246.1 3 - 4 496 55
XM_012959096.2 XP_012814550.1 3 - 4 707 55
GenBank mRNA CR760145.2 - 3 - 4 496 55

View Full Product Details