Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan15
Entrez Gene ID
733836
Gene Name
tetraspanin 15
Gene Aliases
-
Chromosome Location
Chr.8: 59280135 - 59297123 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045589.1 NP_001039054.1 2 - 3 403 62
GenBank mRNA BC155372.1 - 2 - 3 364 62
BC160400.1 - 2 - 3 472 62
CR848522.2 - 2 - 3 403 62

View Full Product Details