Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mxd1
Entrez Gene ID
779675
Gene Name
MAX dimerization protein 1
Gene Aliases
Mad1, mad, xmad1
Chromosome Location
Chr.3: 111906314 - 111908417 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078760.1 NP_001072228.1 - 457 85
GenBank mRNA BC118812.1 - - 457 85

View Full Product Details