Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pex11g
Entrez Gene ID
780227
Gene Name
peroxisomal biogenesis factor 11 gamma
Gene Aliases
-
Chromosome Location
Chr.1: 84269745 - 84274200 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079302.1 NP_001072770.1 2 - 3 410 75
XM_012965877.2 XP_012821331.1 2 - 3 498 75
GenBank mRNA BC125806.1 - 2 - 3 410 75

View Full Product Details