Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mafk
Entrez Gene ID
100036635
Gene Name
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Aliases
nfe2u, p18
Chromosome Location
Chr.9: 23903026 - 23931927 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097194.1 NP_001090663.1 2 - 3 140 99
XM_012969918.2 XP_012825372.1 2 - 3 178 99
XM_012969919.2 XP_012825373.1 3 - 4 274 99
GenBank mRNA BC127302.1 - 2 - 3 140 99

View Full Product Details