Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
p3h1
Entrez Gene ID
100036679
Gene Name
prolyl 3-hydroxylase 1
Gene Aliases
lepre1, leprecan
Chromosome Location
Chr.7: 68525212 - 68553379 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097231.1 NP_001090700.1 3 - 4 837 100
XM_018095197.1 XP_017950686.1 3 - 4 1139 100
GenBank mRNA BC128622.1 - 3 - 4 837 100

View Full Product Details