Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mtfp1
Entrez Gene ID
100124873
Gene Name
mitochondrial fission process 1
Gene Aliases
mtp18
Chromosome Location
Chr.1: 131843383 - 131848892 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102827.1 NP_001096297.1 1 - 2 156 70
GenBank mRNA BC135607.1 - 1 - 2 156 70

View Full Product Details