Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c1orf50
Entrez Gene ID
549419
Gene Name
chromosome 1 open reading frame 50
Gene Aliases
-
Chromosome Location
Chr.7: 68570115 - 68577063 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016665.3 NP_001016665.1 3 - 4 280 92
XM_012966617.2 XP_012822071.1 4 - 5 464 92
GenBank mRNA BC158946.1 - 3 - 4 280 92
BC171198.1 - 3 - 4 271 92
BC171202.1 - 3 - 4 271 92
CR760289.2 - 4 - 5 440 92

View Full Product Details