Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zpr1
Entrez Gene ID
549888
Gene Name
ZPR1 zinc finger
Gene Aliases
znf259
Chromosome Location
Chr.7: 83892565 - 83907449 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017134.3 NP_001017134.1 9 - 10 977 70
GenBank mRNA CR761047.3 - 9 - 10 977 70

View Full Product Details