Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kcnk7
Entrez Gene ID
100144289
Gene Name
potassium channel, two pore domain subfamily K, member 7
Gene Aliases
k2p7.1, twik3
Chromosome Location
Chr.8: 83434236 - 83440997 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122816.1 NP_001116288.1 1 - 2 374 98
GenBank mRNA BC158151.1 - 1 - 2 374 98
CR761128.2 - 1 - 2 385 98

View Full Product Details