Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tulp3
Entrez Gene ID
100144732
Gene Name
tubby like protein 3
Gene Aliases
tubl3
Chromosome Location
Chr.3: 1805683 - 1822359 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123481.1 NP_001116953.1 - 111 58
GenBank mRNA BC160638.1 - - 111 58

View Full Product Details