Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc2a7
Entrez Gene ID
100144999
Gene Name
solute carrier family 2 member 7
Gene Aliases
glut7
Chromosome Location
Chr.1: 141850908 - 141873659 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126561.1 NP_001120033.1 7 - 8 920 79
GenBank mRNA BC158339.1 - 7 - 8 920 79

View Full Product Details