Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100285161
Entrez Gene ID
100285161
Gene Name
ATSWI3A
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001158055.1 NP_001151527.1 - 285 69
GenBank mRNA EU971062.1 - - 285 69

View Full Product Details