Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
fab1
Entrez Gene ID
100273168
Gene Name
fatty acid biosynthesis1
Gene Aliases
ZEAMMB73_382080
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001147614.1 NP_001141086.1 - 772 98
GenBank mRNA BT040366.1 - - 772 98
BT085766.1 - - 738 98
BT088241.1 - - 652 98

View Full Product Details