Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pep7
Entrez Gene ID
542479
Gene Name
phosphoenolpyruvate carboxylase7
Gene Aliases
Ppc1C, ZEAMMB73_218220
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112033.1 NP_001105503.1 - 1405 89
GenBank mRNA AB012228.1 - - 1405 89

View Full Product Details