Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
serk1
Entrez Gene ID
542015
Gene Name
somatic embryogenesis receptor-like kinase1
Gene Aliases
ZEAMMB73_384900
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111662.1 NP_001105132.1 - 94 85
GenBank mRNA AJ277702.1 - - 94 85
BT042243.1 - - 338 85

View Full Product Details