Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pip1b
Entrez Gene ID
541779
Gene Name
plasma membrane intrinsic protein1
Gene Aliases
pip1-2
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111464.1 NP_001104934.1 - 1047 113
GenBank mRNA AF131201.1 - - 1047 113

View Full Product Details