Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
hsp26
Entrez Gene ID
542576
Gene Name
heat shock protein26
Gene Aliases
ZEAMMB73_154671
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112113.1 NP_001105583.1 - 251 71
GenBank mRNA EU951996.1 - - 292 71
EU952790.1 - - 285 71
EU964533.1 - - 285 71
L28712.1 - - 251 71

View Full Product Details