Gene Transcripts
- Gene Symbol
- Speer5-ps1
- Entrez Gene ID
- 70365
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 5, pseudogene 1
- Gene Aliases
- 1700007C10Rik, SPEER-5, Speer5-ps
- Chromosome Location
- Chr. 10: 44170447 - 44219585 on Build GRCm38
- UniGene ID
- Mm.87073
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001582.2 |
|
7 |
1026 |
|
NR_027506.1 |
|
4 |
564 |
|
GenBank mRNA |
DV646842.1 |
- |
4 |
564 |
|
View Full Product Details