Gene Transcripts

Gene Symbol
Snora64
Entrez Gene ID
104366
Gene Name
small nucleolar RNA, H/ACA box 64
Gene Aliases
MBI-29, Rnu64
Chromosome Location
Chr. 17: 24720789 - 24720905 on Build GRCm38
UniGene ID
Mm.157452
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002897.1 1 20
GenBank mRNA AF357417.1 - 1 20


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AV461327.1 - 1 43 Chr. 17 - 24720766 - 24720906 [+]
GenBankmRNA BU516967.1 - 1 675 Chr. 17 - 24720134 - 24721069 [+]
GenBankmRNA BU511787.1 - 1 676 Chr. 17 - 24720133 - 24720973 [+]
GenBankmRNA BM949092.1 - 2 466 Chr. 17 - 24720262 - 24721106 [+]
GenBankmRNA CX217171.1 - 1 522 Chr. 17 - 24720287 - 24721142 [+]
GenBankmRNA BI653619.1 - 2 462 Chr. 17 - 24720281 - 24721170 [+]
GenBankmRNA CN717869.1 - 1 67 Chr. 17 - 24720742 - 24721375 [+]
GenBankmRNA CN671732.1 - 2 458 Chr. 17 - 24720271 - 24720923 [+]
GenBankmRNA CX231626.1 - 2 450 Chr. 17 - 24720278 - 24720949 [+]
GenBankmRNA CX217169.1 - 1 522 Chr. 17 - 24720287 - 24720994 [+]
GenBankmRNA BG806892.1 - 1 135 Chr. 17 - 24720674 - 24721273 [+]
GenBankmRNA BI658732.1 - 2 590 Chr. 17 - 24720135 - 24721095 [+]

View Full Product Details