Gene Transcripts

Gene Symbol
TMPRSS7
Entrez Gene ID
344805
Gene Name
transmembrane protease, serine 7
Gene Aliases
Chromosome Location
Chr. 3: 112039067 - 112081269 on Build GRCh38
UniGene ID
Hs.435490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026734.1 5 572
GenBank mRNA AK131211.1 - 5 572

View Full Product Details