Gene Transcripts

Gene Symbol
SDK1
Entrez Gene ID
221935
Gene Name
sidekick cell adhesion molecule 1
Gene Aliases
Chromosome Location
Chr. 7: 3301448 - 4269000 on Build GRCh38
UniGene ID
Hs.653013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079653.1 NP_001073121.1 19, 20 2789
XM_011515188.1 XP_011513490.1 39, 40 5485
XM_011515189.1 XP_011513491.1 39, 40 5482
GenBank mRNA AL833354.1 - 19, 20 2786

View Full Product Details