Gene Transcripts

Gene Symbol
OR7E91P
Entrez Gene ID
79315
Gene Name
olfactory receptor family 7 subfamily E member 91 pseudogene
Gene Aliases
Chromosome Location
Chr. 2: 71024075 - 71029931 on Build GRCh38
UniGene ID
Hs.327033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002185.3 3 1155
GenBank mRNA BC014374.1 - 3 1158
BC030991.1 - 3 1194

Gene Symbol
OR7E2P
Entrez Gene ID
8587
Gene Name
olfactory receptor family 7 subfamily E member 2 pseudogene
Gene Aliases
OR11-6, OR7E143P, OR7E51P, OR7E52P, OR7F2P, OST245, hg94
Chromosome Location
Chr. 11: 86856931 - 86857954 on Build GRCh38
UniGene ID
Hs.741959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045004.1 1 880

View Full Product Details