Gene Transcripts
- Gene Symbol
- TIPARP-AS1
- Entrez Gene ID
- 100287227
- Gene Name
- TIPARP antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 156673171 - 156675713 on Build GRCh38
- UniGene ID
- Hs.12813
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027954.1 |
|
1 |
2076 |
|
GenBank mRNA |
BC033324.1 |
- |
1 |
2042 |
|
BC038770.1 |
- |
1 |
710 |
|
View Full Product Details