Gene Transcripts
- Gene Symbol
- NAP1L6
- Entrez Gene ID
- 645996
- Gene Name
- nucleosome assembly protein 1 like 6
- Gene Aliases
- Chromosome Location
- Chr. X: 73126037 - 73128080 on Build GRCh38
- UniGene ID
- Hs.617486
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027291.1 |
|
1 |
1647 |
|
GenBank mRNA |
AK090915.1 |
- |
1 |
1650 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM715105.1 |
- |
1 |
285 |
|
Chr. X - 73126037 - 73126718 [-] |
View Full Product Details