Gene Transcripts

Gene Symbol
FAM92A1P2
Entrez Gene ID
403315
Gene Name
family with sequence similarity 92, member A3
Gene Aliases
FAM92A3
Chromosome Location
Chr. 4: 183037665 - 183040119 on Build GRCh38
UniGene ID
Hs.652187
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003612.2 1 1751
GenBank mRNA BC022019.1 - 1 1714
BC047622.1 - 1 1751
BC059396.1 - 1 1751

View Full Product Details