Gene Transcripts
- Gene Symbol
- RACGAP1P
- Entrez Gene ID
- 83956
- Gene Name
- Rac GTPase activating protein 1 pseudogene
- Gene Aliases
- FKSG42
- Chromosome Location
- Chr. 12: 45062618 - 45065411 on Build GRCh38
- UniGene ID
- Hs.650500
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026583.1 |
|
1 |
352 |
|
GenBank mRNA |
BC125190.1 |
- |
1 |
343 |
|
BC125191.1 |
- |
1 |
343 |
|
View Full Product Details