Gene Transcripts
- Gene Symbol
- TDH
- Entrez Gene ID
- 157739
- Gene Name
- L-threonine dehydrogenase (pseudogene)
- Gene Aliases
- SDR14E1P
- Chromosome Location
- Chr. 8: 11339637 - 11368452 on Build GRCh38
- UniGene ID
- Hs.583896
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001578.1 |
|
1, 2 |
87 |
|
GenBank mRNA |
AJ301562.1 |
- |
1, 2 |
87 |
|
View Full Product Details