Gene Transcripts
- Gene Symbol
- SNORA71A
- Entrez Gene ID
- 26777
- Gene Name
- small nucleolar RNA, H/ACA box 71A
- Gene Aliases
- RNU71A, U71a
- Chromosome Location
- Chr. 20: 38427306 - 38427443 on Build GRCh38
- UniGene ID
- Hs.681779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002911.1 |
|
1 |
119 |
|
GenBank mRNA |
Y11165.1 |
- |
1 |
119 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM918074.1 |
- |
|
121 |
|
Chr. - - [-] |
View Full Product Details