Gene Transcripts
- Gene Symbol
- FAM99A
- Entrez Gene ID
- 387742
- Gene Name
- family with sequence similarity 99 member A (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. 11: 1665599 - 1667856 on Build GRCh38
- UniGene ID
- Hs.567908
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026643.1 |
|
3 |
1227 |
|
GenBank mRNA |
AK124823.1 |
- |
1 |
1831 |
|
CR627417.1 |
- |
3 |
1254 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000382167.2 |
- |
3 |
1229 |
|
Chr. 11 - 1665595 - 1667855 [+] |
View Full Product Details