Gene Transcripts
- Gene Symbol
- ANKRD36BP1
- Entrez Gene ID
- 84832
- Gene Name
- ankyrin repeat domain 36B pseudogene 1
- Gene Aliases
- ANKRD26L1, ANKRD36BL1
- Chromosome Location
- Chr. 1: 168245565 - 168247430 on Build GRCh38
- UniGene ID
- Hs.537067
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026844.1 |
|
1 |
1433 |
|
GenBank mRNA |
BC007072.1 |
- |
1 |
1346 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HX784625.1 |
- |
1 |
371 |
|
Chr. 1 - 168245753 - 168246368 [-] |
GenBankmRNA |
HX786161.1 |
- |
1 |
371 |
|
Chr. 1 - 168245753 - 168246368 [-] |
View Full Product Details