Gene Transcripts
- Gene Symbol
- SNORA2A
- Entrez Gene ID
- 677793
- Gene Name
- small nucleolar RNA, H/ACA box 2A
- Gene Aliases
- ACA2A
- Chromosome Location
- Chr. 12: 48656648 - 48656782 on Build GRCh38
- UniGene ID
- Hs.689723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002950.1 |
|
1 |
93 |
|
GenBank mRNA |
AJ609426.1 |
- |
1 |
93 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CV322587.1 |
- |
1 |
146 |
|
Chr. 12 - 48656613 - 48656835 [-] |
View Full Product Details