Gene Transcripts
- Gene Symbol
- SNORA71E
- Entrez Gene ID
- 677821
- Gene Name
- small nucleolar RNA, H/ACA box 71E
- Gene Aliases
- ACA39, SNORA39
- Chromosome Location
- Chr. 20: 38448083 - 38448218 on Build GRCh38
- UniGene ID
- Hs.741059
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002972.1 |
|
1 |
83 |
|
GenBank mRNA |
AJ609475.1 |
- |
1 |
83 |
|
- Gene Symbol
- SNHG11
- Entrez Gene ID
- 128439
- Gene Name
- small nucleolar RNA host gene 11
- Gene Aliases
- C20orf198, LINC00101, NCRNA00101
- Chromosome Location
- Chr. 20: 38446654 - 38450921 on Build GRCh38
- UniGene ID
- Hs.16936
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055434.1 |
- |
1 |
1513 |
|
- Gene Symbol
- SNORA60
- Entrez Gene ID
- 677837
- Gene Name
- small nucleolar RNA, H/ACA box 60
- Gene Aliases
- ACA60
- Chromosome Location
- Chr. 20: 38449369 - 38449504 on Build GRCh38
- UniGene ID
- Hs.16936
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091209.1 |
- |
1 |
330 |
|
View Full Product Details