Gene Transcripts
- Gene Symbol
- SNORD9
- Entrez Gene ID
- 692053
- Gene Name
- small nucleolar RNA, C/D box 9
- Gene Aliases
- mgU6-53B
- Chromosome Location
- Chr. 14: 21392151 - 21392253 on Build GRCh38
- UniGene ID
- Hs.675950
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003029.2 |
|
1 |
67 |
|
View Full Product Details