Gene Transcripts
- Gene Symbol
- SNORD13
- Entrez Gene ID
- 692084
- Gene Name
- small nucleolar RNA, C/D box 13
- Gene Aliases
- RNU13, SNORD13A, U13
- Chromosome Location
- Chr. 8: 33513475 - 33513578 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003041.1 |
|
1 |
29 |
|
- Gene Symbol
- SNORD13P1
- Entrez Gene ID
- 6076
- Gene Name
- small nucleolar RNA, C/D box 13 pseudogene 1
- Gene Aliases
- RNU13P1, U13, U13.32A
- Chromosome Location
- Chr. 22: 42076059 - 42076161 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X58061.1 |
- |
1 |
80 |
|
View Full Product Details