Gene Transcripts
- Gene Symbol
- SNORD67
- Entrez Gene ID
- 692108
- Gene Name
- small nucleolar RNA, C/D box 67
- Gene Aliases
- HBII-166
- Chromosome Location
- Chr. 11: 46762389 - 46762499 on Build GRCh38
- UniGene ID
- Hs.693252
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003056.1 |
|
1 |
71 |
|
View Full Product Details