Gene Transcripts
- Gene Symbol
- NUDT9P1
- Entrez Gene ID
- 119369
- Gene Name
- nudix hydrolase 9 pseudogene 1
- Gene Aliases
- C10orf98, bA56M3.1
- Chromosome Location
- Chr. 10: 91151995 - 91153083 on Build GRCh38
- UniGene ID
- Hs.441287
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002779.1 |
|
1 |
817 |
|
GenBank mRNA |
BC029544.1 |
- |
1 |
817 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY192208.1 |
- |
1 |
191 |
|
Chr. 10 - 91151957 - 91152425 [-] |
View Full Product Details