Gene Transcripts
- Gene Symbol
- PWRN2
- Entrez Gene ID
- 791115
- Gene Name
- Prader-Willi region non-protein coding RNA 2
- Gene Aliases
- NCRNA00199
- Chromosome Location
- Chr. 15: 24164779 - 24169906 on Build GRCh38
- UniGene ID
- Hs.560378
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026647.1 |
|
1 |
4816 |
|
GenBank mRNA |
BC032951.1 |
- |
2 |
3942 |
|
BC070120.1 |
- |
2 |
3937 |
|
View Full Product Details