Gene Transcripts

Gene Symbol
FAM86EP
Entrez Gene ID
348926
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
Chromosome Location
Chr. 4: 3941760 - 3955428 on Build GRCh38
UniGene ID
Hs.591720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_130740.1 4 2233
NR_130741.1 4 1287
NR_130742.1 4 1237

View Full Product Details