Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003561.2 | 5, 6 | 1921 | ||
NR_027768.1 | 3, 4 | 2216 | |||
GenBank mRNA | AK131353.1 | - | 5, 6 | 1914 | |
AL834175.1 | - | 3, 4 | 1969 | ||
BC150309.1 | - | 3, 4 | 1677 | ||
BX648212.1 | - | 4, 5 | 2198 |