Gene Transcripts
- Gene Symbol
- SNORA27
- Entrez Gene ID
- 619499
- Gene Name
- small nucleolar RNA, H/ACA box 27
- Gene Aliases
- ACA27
- Chromosome Location
- Chr. 13: 27255401 - 27255526 on Build GRCh38
- UniGene ID
- Hs.689719
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002575.1 |
|
1 |
115 |
|
GenBank mRNA |
AJ609439.1 |
- |
1 |
115 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA914688.1 |
- |
|
70 |
|
Chr. 6 - 73519144 - 73521040 [-] |
GenBankmRNA |
BG111952.1 |
- |
1 |
571 |
|
Chr. 13 - 27254945 - 27255754 [+] |
GenBankmRNA |
BG529829.1 |
- |
1 |
324 |
|
Chr. 13 - 27255192 - 27256052 [+] |
View Full Product Details