Gene Transcripts

Gene Symbol
HCCAT5
Entrez Gene ID
283902
Gene Name
hepatocellular carcinoma associated transcript 5 (non-protein coding)
Gene Aliases
FJ222407, HTA
Chromosome Location
Chr. 16: 73092349 - 73093774 on Build GRCh38
UniGene ID
Hs.382776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027756.1 3 1040
GenBank mRNA BC019009.2 - 3 764
FJ222407.1 - 3 1040
FJ222408.1 - 1 1387

View Full Product Details