Gene Transcripts
- Gene Symbol
- LOC107986705
- Entrez Gene ID
- 107986705
- Gene Name
- integrator complex subunit 4-like
- Gene Aliases
- Chromosome Location
- Chr. 7: 65191623 - 65214524 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- INTS4P2
- Entrez Gene ID
- 644619
- Gene Name
- integrator complex subunit 4 pseudogene 2
- Gene Aliases
- INTS4L2
- Chromosome Location
- Chr. 7: 65647854 - 65718688 on Build GRCh38
- UniGene ID
- Hs.676517
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027392.2 |
|
13 |
1645 |
|
View Full Product Details