Gene Transcripts
- Gene Symbol
- DLEU2L
- Entrez Gene ID
- 79469
- Gene Name
- deleted in lymphocytic leukemia 2-like
- Gene Aliases
- BCMSUNL
- Chromosome Location
- Chr. 1: 63548980 - 63550636 on Build GRCh38
- UniGene ID
- Hs.659291
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002771.1 |
|
1 |
1622 |
|
GenBank mRNA |
AF254117.1 |
- |
1 |
1622 |
|
View Full Product Details