Gene Transcripts

Gene Symbol
LINC00483
Entrez Gene ID
55018
Gene Name
long intergenic non-protein coding RNA 483
Gene Aliases
C17orf73
Chromosome Location
Chr. 17: 50761034 - 50767515 on Build GRCh38
UniGene ID
Hs.389460
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_073199.1 4 460
GenBank mRNA AK000670.1 - 4 461
AK000701.1 - 3 2395
BP382918.1 - 4 460

View Full Product Details