Gene Transcripts
- Gene Symbol
- LINC00483
- Entrez Gene ID
- 55018
- Gene Name
- long intergenic non-protein coding RNA 483
- Gene Aliases
- C17orf73
- Chromosome Location
- Chr. 17: 50761034 - 50767515 on Build GRCh38
- UniGene ID
- Hs.389460
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073199.1 |
|
4 |
399 |
|
GenBank mRNA |
AK000670.1 |
- |
4 |
400 |
|
AK000701.1 |
- |
3 |
2334 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000300458.1 |
- |
3 |
2332 |
|
Chr. 17 - 50761029 - 50767518 [-] |
View Full Product Details