Gene Transcripts
- Gene Symbol
- FAM86FP
- Entrez Gene ID
- 653113
- Gene Name
- family with sequence similarity 86, member A pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 12: 8231049 - 8242946 on Build GRCh38
- UniGene ID
- Hs.661749
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024254.1 |
|
5 |
2111 |
|
View Full Product Details