Gene Transcripts

Gene Symbol
TSPY26P
Entrez Gene ID
128854
Gene Name
testis specific protein, Y-linked 26, pseudogene
Gene Aliases
TSPYL3, bA392M18.1
Chromosome Location
Chr. 20: 32189146 - 32190360 on Build GRCh38
UniGene ID
Hs.647447
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002781.1 1 1179
GenBank mRNA BC101556.1 - 1 1179
BC101558.1 - 1 1179


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN430810.1 - 1 336 Chr. 20 - 32188777 - 32189517 [-]
GenBankmRNA AL935106.1 - 1 374 Chr. 20 - 32188836 - 32189554 [-]

View Full Product Details