Gene Transcripts
- Gene Symbol
- SNORA12
- Entrez Gene ID
- 677800
- Gene Name
- small nucleolar RNA, H/ACA box 12
- Gene Aliases
- U108
- Chromosome Location
- Chr. 10: 100237156 - 100237302 on Build GRCh38
- UniGene ID
- Hs.662165
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002954.1 |
|
1 |
89 |
|
GenBank mRNA |
AM055730.1 |
- |
1 |
89 |
|
View Full Product Details