Gene Transcripts
- Gene Symbol
- SNORA30
- Entrez Gene ID
- 677813
- Gene Name
- small nucleolar RNA, H/ACA box 30
- Gene Aliases
- ACA30, SNORA30A
- Chromosome Location
- Chr. 16: 30710537 - 30710665 on Build GRCh38
- UniGene ID
- Hs.689718
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002966.1 |
|
1 |
70 |
|
GenBank mRNA |
AJ609440.1 |
- |
1 |
70 |
|
View Full Product Details